missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, APC-R700, Clone: RPA-T4, BD Horizon™
Mouse Monoclonal Antibody
2165.00 NOK - 4415.00 NOK
Specifications
Antigen | CD4 |
---|---|
Clone | RPA-T4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-R700 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15863999
|
BD Biosciences
564976 |
25 Tests |
2165.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15853999
|
BD Biosciences
564975 |
100 Tests |
4415.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.Specifications
CD4 | |
Flow Cytometry | |
APC-R700 | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
IgG1 Îş | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. |
RPA-T4 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
L3T4; T-cell surface antigen T4/Leu-3; W3/25; CD4 antigen (p55) | |
Primary | |
Vol. Per test: 5ÎĽL |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD4 Mouse anti-Human, APC-R700, Clone: RPA-T4, BD Horizon™