missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, APC-R700, Clone: RPA-T4, BD Horizon™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 564976

2163.54 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15863999

  • 2165.00 NOK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD4
Monoclonal
APC-R700
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Mouse
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
IgG1 κ
Flow Cytometry
RPA-T4
Vol. Per test: 5μL
L3T4; T-cell surface antigen T4/Leu-3; W3/25; CD4 antigen (p55)
25 Tests
Cell Biology
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD4 Mouse anti-Human, APC-R700, Clone: RPA-T4, BD Horizon™ > 25 tests; APC-R700

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.