missing translation for 'onlineSavingsMsg'
Learn More

CD278 (ICOS) Monoclonal Antibody (C398.4A), Functional Grade, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

3070.00 NOK - 150400.00 NOK

Specifications

Antigen CD278 (ICOS)
Clone C398.4A
Concentration 1 mg/mL
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15238847
View Documents
Invitrogen
16-9949-82
100 μg
3070.00 NOK
100µg
Please to purchase this item. Need a web account? Register with us today!
17171632
View Documents
Affymetrix eBioscience
16-9949-025
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17181632
View Documents
Affymetrix eBioscience
16-9949-050
50 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The C398.4A MAb reacts with ICOS (Inducible Costimulatory molecule), also known as H4/CRP-1/AILIM, and cross-reacts with mouse and rat ICOS. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. A dimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1, is thought to play a protective role in inflammatory autoimmune diseases and be involved in the development of Th2 cells. C398.4A is reported to display strong costimulation of proliferation of T cell clones, intermediate costimulation on activated T cells, and weak costimulation on fresh resting T cells. It is a good costimulator of IL-10 production especially from pre-activated cells that express high level of H4/ICOS. The epitope recognized by C398.4A is different from that bound by the ICOSL (i.e., C398.4A does not block binding of ICOSL).

ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications

Specifications

CD278 (ICOS)
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115
IgG
Affinity chromatography
Antibody
C398.4A
Flow Cytometry, Functional Assay
Functional Grade
Armenian Hamster
Mouse, Rat, Rhesus Monkey
Q9R1T7, Q9WVS0
54167, 64545, 705788
Primary
4° C
ICOS
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD278 (ICOS) Monoclonal Antibody (C398.4A), Functional Grade, eBioscience™, Invitrogen™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.