Learn More
CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
14440.00 NOK - 36070.00 NOK
Specifications
Antigen | CD157 |
---|---|
Clone | eBioSY11B5 (SY11B5) |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17100582
|
Invitrogen
14-1579-37 |
2 mg |
14440.00 NOK
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17110582
|
Affymetrix eBioscience
14-1579-95 |
10 mg | N/A | ||||||
Description
The eBioSY11B5 MAb recognizes human CD157 (Mo5, BST-1). CD157 is a 42-45 kDa, GPI-anchored protein with structural and functional similarities with CD38. CD157 was initially cloned because of its expression on monocytes and macrophages, and was subsequently discovered to be the same protein named BST-1, discovered for its expression on bone marrow stromal cells and its ability to stimulate the proliferation of a mouse pre-B cell line. CD157 is a pleiotropic ectoenzyme and is thought to act independently as an enzyme and receptor. Similar to CD38, CD157 is involved in the metabolism of NAD+ and this activity may be involved in regulating intracellular Ca2+ levels. As a receptor, upon binding of its putative ligand, CD157 is thought to initiate a signal transduction cascade resulting in the phosphorylation of cytoplasmic proteins including focal adhesion kinase (FAK). The mechanism and functional significance of CD157-initiated signal transduction remain to be fully characterized.
Bone marrow stromal cell antigen 1 (BST1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-B cell growth. BST1, which is also known as ADP-rybosyl cyclase 2 and DAMP-1, is an enzyme which is expressed by BST1 gene in human. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.Specifications
CD157 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
683 | |
Primary | |
4° C | |
Bst1 |
eBioSY11B5 (SY11B5) | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q10588 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.