missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133-1,2,4,7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CD133-1,2,4,7 |
---|---|
Concentration | 167 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899414
|
Proteintech
19945-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889414
|
Proteintech
19945-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD133-1, 2, 4, 7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human CD133-1, 2, 4, 7. This antibody recognizes human, mouse antigen. The CD133-1, 2, 4, 7 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
CD133-1,2,4,7 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
167 μg/mL | |
Polyclonal | |
Rabbit | |
8842 | |
Primary |