Learn More
CD133-1,2,4,7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19945-1-AP-20UL
 View more versions of this product
Product Code. 16899414
  • 1611.84 NOK / 20µL
Explore more special offers
Description
Description
The CD133-1, 2, 4, 7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human CD133-1, 2, 4, 7. This antibody recognizes human, mouse antigen. The CD133-1, 2, 4, 7 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CD133-1,2,4,7
Polyclonal
Unconjugated
20 μL
8842
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
167 μg/mL
Rabbit
Primary
Human, Mouse