missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC59 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26387-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of the transcription complexes of the pulmonary surfactant-associated protein-B (SFTPB) and -C (SFTPC). Enhances homeobox protein Nkx-2.1-activated SFTPB and SFTPC promoter activities.Specifications
CCDC59 | |
Polyclonal | |
Unconjugated | |
CCDC59 | |
BR22, CCDC59, HSPC128, TAP26, TTF 1 associated protein 26, TTF 1 associated protein BR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29080 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P031 | |
CCDC59 | |
CCDC59 Fusion Protein Ag23852 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction