missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC59 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CCDC59 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16804875
|
Proteintech
26387-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16894865
|
Proteintech
26387-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Component of the transcription complexes of the pulmonary surfactant-associated protein-B (SFTPB) and -C (SFTPC). Enhances homeobox protein Nkx-2.1-activated SFTPB and SFTPC promoter activities.Specifications
CCDC59 | |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q9P031 | |
29080 | |
CCDC59 Fusion Protein Ag23852 | |
Primary | |
-20°C | |
CCDC59 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BR22, CCDC59, HSPC128, TAP26, TTF 1 associated protein 26, TTF 1 associated protein BR2 | |
CCDC59 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title