missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ FcXP Pre-packed Column

Thermo Scientific™ CaptureSelect™ FcXP Affinity Matrix is offered here in a pre-packed format with connectors that are compatible with standard chromatography equipment.

16830.00 NOK

Specifications

Binding Capacity 40 to 50 g IgG/L resin
Flow Rate 150 to 300 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide-activated
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18406428
View Documents
Thermo Scientific™
494371205
1 x 5 mL Column
16830.00 NOK
5mL
Please to purchase this item. Need a web account? Register with us today!
18486468
View Documents
Thermo Scientific™
494371201
5 x 1 mL Columns
16830.00 NOK
5mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaptureSelect FcXP Affinity Matrix is offered here in a pre-packed format with connectors that are compatible with standard chromatography equipment.

CaptureSelect FcXP Affinity Matrix is the successor to CaptureSelect FcXL Affinity Matrix, specifically designed for higher binding capacity. The selectivity is identical; the ligand recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4). But the matrix features an increased binding capacity (>40 g IgG/L), while maintaining the elution characteristics of CaptureSelect FcXL Affinity Matrix.

Features of this affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • Human specific, no binding to bovine antibodies
  • Excellent scalability
  • Non-animal derived

CaptureSelect FcXP pre-packed columns can be used for the purification of IgG and Fc fusion proteins directly from complex source materials and enable a robust and efficient purification process with excellent purity in one step.

Characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 ÎĽm
  • Ligand: CaptureSelect FcXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 40–50 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4, or 100 mM Tris, 2.0 M MgCl2, 40% (v/v) propylene glycol, pH 7
  • Flow characteristics: 150–300 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Specifications

Specifications

40 to 50 g IgG/L resin
Agarose-based, epoxide-activated
Wet Ice
CaptureSelect™
FcXP affinity ligand
150 to 300 cm/hr. (Up to 2 bar)
Store at 2°C to 8°C.
65 ±10 μm
Pre-packed Column
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CaptureSelect™ FcXP Pre-packed Column

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.