Learn More
BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories
Allows the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.Dispensette™III, Dispensette™ Organic Remote Dispenser Parts and Accessories include threaded adapters, a table/shelf clamp and a support rod for the wall mounting unit.
324.00 NOK - 749.00 NOK
Specifications
For Use With (Equipment) | Dispensette™III, Dispensette™ Organic |
---|---|
Quantity | 1 Ea. |
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
10613012
|
BRAND™
704268 |
Support Rod Connector for Wall Mounting Unit |
749.00 NOK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
10643202
|
BRAND™
704272 |
Table/Shelf Clamp for Wall Mounting Unit |
324.00 NOK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Description
- Dispensette™ parts and accessories allow the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.
- Dispenses accurate volumes directly from drums and bulk refills.
- A filter in the drum adapter minimizes risk of contaminating high-purity reagents.
- A quick-release connector with integrated valves allows quick changing of the bulk container.
- The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
- Maximum delivery height is approximately 1.2 m
Specifications
Dispensette™III, Dispensette™ Organic | |
1 Ea. |
Product Title
BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories