Learn More
BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories
Allows the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.Dispensette™III, Dispensette™ Organic Remote Dispenser Parts and Accessories include threaded adapters, a table/shelf clamp and a support rod for the wall mounting unit.
Brand: BRAND™ 704268
500.12 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Dispensette™ parts and accessories allow the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.
- Dispenses accurate volumes directly from drums and bulk refills.
- A filter in the drum adapter minimizes risk of contaminating high-purity reagents.
- A quick-release connector with integrated valves allows quick changing of the bulk container.
- The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
- Maximum delivery height is approximately 1.2 m
Specifications
Support Rod Connector for Wall Mounting Unit | |
Dispensette™III, Dispensette™ Organic | |
1 Ea. |