Learn More
GPE Scientific™ Borosilicate Glass Stirrer
For use with overhead stirrer motors
1250.00 NOK - 1280.00 NOK
Specifications
For Use With (Equipment) | Overhead Stirrer Motors |
---|---|
Type | Uni-Form Precision Glass Stirrers |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
12977066
|
GPE Scientific™
30502 |
B24 |
1280.00 NOK
Each |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
11786843
|
GPE Scientific™
30503 |
B29 |
1280.00 NOK
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
12967066
|
GPE Scientific™
30501 |
B19 |
1250.00 NOK
Each |
Estimated Shipment: 04-03-2025
Log in to see stock. |
|||||
Description
- For use with overhead stirrer motors
- Non-whip and autoclavable
- The PTFE blades are easily removed or replaced
Specifications
Overhead Stirrer Motors | |
Uni-Form Precision Glass Stirrers |