Learn More
GPE Scientific™ Borosilicate Glass Stirrer
For use with overhead stirrer motors
Brand: GPE Scientific™ 30501
This item is not returnable.
View return policy
Description
- For use with overhead stirrer motors
- Non-whip and autoclavable
- The PTFE blades are easily removed or replaced
Specifications
Overhead Stirrer Motors | |
B19 | |
Uni-Form Precision Glass Stirrers | |
Borosilicate Glass |