Learn More
Bohlender™ BOLA™ Balls
Balls for use as splash guards or extending surfaces.
Brand:  Bohlender™ H964-15
 View more versions of this product
Product Code. 17157424
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made of solid PTFE, with smooth surface
  • Different packing units
  • For extension of surfaces such as distillation apparatus or as splash guard
Specifications
Specifications
PTFE
15 mm
Smooth Surface
50 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bohlender™ BOLA™ Balls > Balls; Diameter (Metric): 15 mm; Material: PTFE; Quantity: 50 Pk.; Description: Smooth Surface