Learn More
Bohlender™ BOLA™ Balls
Balls for use as splash guards or extending surfaces.
Brand:  Bohlender™ H964-09
 View more versions of this product
Product Code. 17147424
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made of solid PTFE, with smooth surface
  • Different packing units
  • For extension of surfaces such as distillation apparatus or as splash guard
Specifications
Specifications
PTFE
9 mm
Smooth Surface
100 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bohlender™ BOLA™ Balls > Balls; Diameter (Metric): 9 mm; Material: PTFE; Quantity: 100 Pk.; Description: Smooth Surface