missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Biotin Control Diet Powder

Biotin Control Diet. Contains egg white spray, corn starch, and salt mixture and vitamin fortification.

8795.00 NOK - 33590.00 NOK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Powder
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11421472
View Documents
MP Biomedicals
0296040910
10 kg
8795.00 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11431472
View Documents
MP Biomedicals
0296040920
20 kg
15485.00 NOK
20kg
Please to purchase this item. Need a web account? Register with us today!
11441472
View Documents
MP Biomedicals
0296040950
50 kg
33590.00 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Biotin Control Diet
  • Contains egg white spray, corn starch, and salt mixture and vitamin fortification
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Powder
Refrigerated
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.