missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Biotin Control Diet Powder

Biotin Control Diet. Contains egg white spray, corn starch, and salt mixture and vitamin fortification.

Brand:  MP Biomedicals 0296040920

 View more versions of this product

Product Code. 11431472

  • 15485.00 NOK / 20kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Biotin Control Diet
  • Contains egg white spray, corn starch, and salt mixture and vitamin fortification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
20 kg
Refrigerated
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.