Learn More
Thermo Scientific Chemicals BICINE, 0.5M buffer soln., pH 7.5
1144.80 NOK - 1796.40 NOK
Chemical Identifiers
CAS | 150-25-4 |
---|---|
Molecular Formula | C6H12NNaO4 |
Molecular Weight (g/mol) | 185.16 |
MDL Number | MFCD00004295 |
InChI Key | MFBDBXAVPLFMNJ-UHFFFAOYSA-M |
PubChem CID | 8761 |
ChEBI | CHEBI:40957 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]acetic acid |
SMILES | [Na+].OCCN(CCO)CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433269
|
Thermo Scientific Chemicals
J60494.AE |
100 mL |
1144.80 NOK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443269
|
Thermo Scientific Chemicals
J60494.AK |
250 mL |
1796.40 NOK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA zwitterionic amino acid buffer agent used for low temperature work. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.
Solubility
Soluble in water (1M) at 20°C, ethanol (slightly), ether (slightly), and chloroform (slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
150-25-4 | |
185.16 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
CHEBI:40957 | |
[Na+].OCCN(CCO)CC([O-])=O |
C6H12NNaO4 | |
MFCD00004295 | |
8761 | |
2-[bis(2-hydroxyethyl)amino]acetic acid |
Specifications
150-25-4 | |
7.5 | |
C6H12NNaO4 | |
Soluble in water (1M) at 20°C,ethanol (slightly),ether (slightly),and chloroform (slightly). | |
[Na+].OCCN(CCO)CC([O-])=O | |
185.16 | |
CHEBI:40957 |
0.5 M | |
Liquid | |
MFCD00004295 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
2-[bis(2-hydroxyethyl)amino]acetic acid | |
8761 | |
BICINE buffer soln. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only