missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BFSP1 (Human) Recombinant Protein
Human BFSP1 full-length ORF ( AAH41483, 1 a.a. - 540 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH41483 |
---|---|
Gene ID (Entrez) | 631 |
Name | Beaded filament structural protein 1, filensin |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16122221
|
Abnova™
H00000631-P01.10ug |
10 μg |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16132221
|
Abnova™
H00000631-P01.25ug |
25 μg |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: MYENECECQLLLKEMLERLNKEADEALLHNLRLQLEAQFLQDDISAAKDRHKKNLLEVQTYISILQQIIHTTPPASIVTSGMREEKLLTEREVAALRSQLEEGREVLSHLQAQRVELQAQTTTLEQAIKSAHECYDDEIQLYNEQIETLRKEIEETERVLEKSSYDCRQLAVAQQTLKNELDRYHRIIEIEGNRLTSAFIETPIPLFTQSHGVSLSTGSGGKDLTRALQDITAAKPRQKALPKNVPRRKEIITKDKTNGALEDAPLKGLEDTKLVQVVLKEESESKFESESKEVSPLTQEGAPEDVPDGGQISKGFGKLYRKVKEKVRSPKEPETPTELYTKERHVLVTGDANYVDPRFYVSSITAKGGVAVSVAEDSVLYDGQVGPSPESPKPPLENGQVGLQEKEDGQPIDQQPIDKEIEPDGAELEGPEEKREGEERDEESRRPCAMVTPGAEEPSIPEPPKPAADQDGAEVLGTRSRSLPEKGPPKALAYKTVEVVESIEKISTESIQTYEETAVIVETMIGKTKSDKKKSGEKSS
Specifications
AAH41483 | |
Beaded filament structural protein 1, filensin | |
125% SDS-PAGE Stained with Coomassie Blue | |
CP115, CP94, FILENSIN, LIFL-H | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
631 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BFSP1 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein