missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BFSP1 (Human) Recombinant Protein

Human BFSP1 full-length ORF ( AAH41483, 1 a.a. - 540 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000631-P01.25ug

 View more versions of this product

Product Code. 16132221

  • 6245.00 NOK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sequence: MYENECECQLLLKEMLERLNKEADEALLHNLRLQLEAQFLQDDISAAKDRHKKNLLEVQTYISILQQIIHTTPPASIVTSGMREEKLLTEREVAALRSQLEEGREVLSHLQAQRVELQAQTTTLEQAIKSAHECYDDEIQLYNEQIETLRKEIEETERVLEKSSYDCRQLAVAQQTLKNELDRYHRIIEIEGNRLTSAFIETPIPLFTQSHGVSLSTGSGGKDLTRALQDITAAKPRQKALPKNVPRRKEIITKDKTNGALEDAPLKGLEDTKLVQVVLKEESESKFESESKEVSPLTQEGAPEDVPDGGQISKGFGKLYRKVKEKVRSPKEPETPTELYTKERHVLVTGDANYVDPRFYVSSITAKGGVAVSVAEDSVLYDGQVGPSPESPKPPLENGQVGLQEKEDGQPIDQQPIDKEIEPDGAELEGPEEKREGEERDEESRRPCAMVTPGAEEPSIPEPPKPAADQDGAEVLGTRSRSLPEKGPPKALAYKTVEVVESIEKISTESIQTYEETAVIVETMIGKTKSDKKKSGEKSS
Specifications

Specifications

AAH41483
Beaded filament structural protein 1, filensin
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BFSP1
GST
631
Wheat germ expression system
25 μg
CP115, CP94, FILENSIN, LIFL-H
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers