missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Agar Slants Prepared Media
Used for the isolation and cultivation of fastidious and nonfastidious microorganisms, including anaerobic and aerobic bacteria
600.00 NOK
Specifications
Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
---|---|
Color | Beige |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Trypticase Soy Agar |
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Liquid | |
Trypticase Soy Agar |
Beige | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
Safety and Handling
missing translation for 'shelfLife' : 548 Days
For in vitro diagnostic use.