missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Agar Slants Prepared Media
Used for the isolation and cultivation of fastidious and nonfastidious microorganisms, including anaerobic and aerobic bacteria
Brand: BD 221086
This item is not returnable.
View return policy
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
15°C to 30°C | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Tube |
Beige | |
Liquid | |
Trypticase Soy Agar | |
Isolation and cultivation of fastidious as well as non-fastidious microorganisms, including anaerobic as well as aerobic bacteria, although it is not the medium of choice for anaerobes | |
10/Pk. |
For in vitro diagnostic use.