Learn More
Brady™ BBP33 Conformable Polyester Thermal Transfer Labels
Designed for high adhesion to textured metals and low surface energy plastics. Brady™ BBP33 Conformable Polyester Thermal Transfer Labels are specifically designed to be pliable and conforming.
Brand: Brady™ 133786
This item is not returnable.
View return policy
Description
- Available in various sizes
- Ideal for marking backer board, DIN rail, relays, PLCs, switches, solenoids, drive motors and other components inside panels
- Flexible with high-tack adhesives for adhesion to wide variety of surfaces and components
Specifications
UL Recognized | |
White | |
Asset Tracking Label | |
5000/Roll | |
0.9 in. |
Rubber, Permanent | |
Gloss | |
BBP33 Printer | |
0.5 in. |