Learn More
MP Biomedicals™ Atherogenic Diet
Semi-purified atherogenic diet modified for mice.
Brand: MP Biomedicals™ 0296040420
This item is not returnable.
View return policy
Description
- Atherogenic diet is rich in cholesterol and other atherogenic factors
- Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
Specifications
Animal Diet | |
Rodent diet | |
20 kg | |
2°C to 8°C |
Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.