Learn More
MP Biomedicals™ Atherogenic Diet
Semi-purified atherogenic diet modified for mice.
Brand:  MP Biomedicals™ 0296040420
 View more versions of this product
Product Code. 11481452
  • 13530.00 NOK / 20kg
Explore more special offers
Description
Description
  • Atherogenic diet is rich in cholesterol and other atherogenic factors
  • Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Rodent diet
20 kg
2°C to 8°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Atherogenic diet is rich in cholesterol and other atherogenic factors. Feeding the mice with atherogenic diet can induce the formation of plaques in the inner lining of arteries and is associated with coronary heart disease.