missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S (Sodium Sulfonate), MP Biomedicals™

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage

1325.00 NOK - 12945.00 NOK

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym Sodium Alizarinsulfonate, Alizarin sodium sulfonate
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443060
View Documents
MP Biomedicals
0210037525
25 g
1325.00 NOK
25g
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
11453060
View Documents
MP Biomedicals
0210037580
100 g
3330.00 NOK
100g
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
11463060
View Documents
MP Biomedicals
0210037590
500 g
12945.00 NOK
500g
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

130-22-3
342.253
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
3955344
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
C14H7NaO7S
MFCD00013049
Sodium Alizarinsulfonate, Alizarin sodium sulfonate
CHEBI:87358
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

130-22-3
MFCD00013049
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
3955344
342.3
Powder
C14H7NaO7S
Sodium Alizarinsulfonate, Alizarin sodium sulfonate
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
342.253
CHEBI:87358
Orange
Alizarin Red S, Indicator: pH Range 3.7 - 5.2
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-981-8

Recommended Storage : Store at Room Temperature (15-30°C).

Product Identifier
  • Alizarin Red S

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Alizarin Red S

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.