missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Red S (Sodium Sulfonate), MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
Formula Weight | 342.3 |
Color | Orange |
Quantity | 100 g |
Physical Form | Powder |
Chemical Name or Material | Alizarin Red S, Indicator: pH Range 3.7 - 5.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alizarin Red S (Sodium Sulfonate), MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction