Learn More
OHAUS™ Adjustable Platform
OHAUS™ Adjustable Platform constructed of stainless steel for durability.
5165.00 NOK - 9775.00 NOK
Product Code | Brand | Height (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15732539
|
OHAUS™
30400121 |
297 mm | 29.70 cm |
5165.00 NOK
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
15702489
|
OHAUS™
30400068 |
460 mm | 45.70 cm |
6720.00 NOK
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||
15712489
|
OHAUS™
30400069 |
610 mm | 61.00 cm |
9775.00 NOK
Each |
In Stock
Log in to see stock. |
||||
Description
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Include the hardware required to mount to shakers
Specifications
Adjustable Platform | |
Stainless Steel |