Learn More
OHAUS™ Adjustable Platform
OHAUS™ Adjustable Platform constructed of stainless steel for durability.
Brand:  OHAUS™ 30400068
 View more versions of this product
Product Code. 15702489
  • 6720.00 NOK / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Description
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Include the hardware required to mount to shakers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adjustable Platform
18.00 in.
45.70 x 45.70 cm
18.00 in.
18.00 in.
Adjustable Platform, 46 X 46 cm
Stainless Steel
45.70 cm
460 mm
45.70 cm
45.70 cm