Learn More
Cytiva Acrylic Plastic Thermostatic Jackets
Brand: Cytiva 18-0011-72
This item is not returnable.
View return policy
Description
Specifications
XK 16/20 Columns | |
XK and K 9 Series Thermostatic Jackets | |
1 Pack |
Acrylic Plastic | |
60°C | |
200 mm |