Learn More
Cytiva Acrylic Plastic Thermostatic Jackets
Brand:  Cytiva 18-0011-75
 View more versions of this product
Product Code. 10691521
  • 2711.00 NOK / Each
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Specifications
XK 16/100 Columns
XK and K 9 Series Thermostatic Jackets
1 Pack
Acrylic Plastic
60°C
1000 mm