Learn More
4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™
4757.00 NOK - 15785.00 NOK
Chemical Identifiers
CAS | 851334-92-4 |
---|---|
Molecular Formula | C15H20BFO4 |
Molecular Weight (g/mol) | 294.129 |
MDL Number | MFCD18383988 |
InChI Key | ZXSGWPHDXQZIAF-UHFFFAOYSA-N |
Synonym | ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoateShow More |
PubChem CID | 23134230 |
IUPAC Name | ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
851334-92-4 | |
294.129 | |
ZXSGWPHDXQZIAF-UHFFFAOYSA-N | |
23134230 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F |
C15H20BFO4 | |
MFCD18383988 | |
ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoateShow More | |
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
Specifications
851334-92-4 | |
MFCD18383988 | |
ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F | |
294.129 | |
294.13 | |
4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester |
C15H20BFO4 | |
14562502 | |
ZXSGWPHDXQZIAF-UHFFFAOYSA-N | |
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate | |
23134230 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.