missing translation for 'onlineSavingsMsg'
Learn More
4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™ Product Code.: 15441916

4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15441916
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15441916

Brand: Thermo Scientific Alfa Aesar H55087.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851334-92-4
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.129
MDL Number MFCD18383988
InChI Key ZXSGWPHDXQZIAF-UHFFFAOYSA-N
Synonym ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 23134230
IUPAC Name ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F

Specifications

Quantity 1 g
Beilstein 14562502
Formula Weight 294.13
Percent Purity 97%
Chemical Name or Material 4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.