missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylhippuric acid, 97%, Thermo Scientific Chemicals
532.80 NOK - 4666.00 NOK
Chemical Identifiers
CAS | 27115-49-7 |
---|---|
Molecular Formula | C10H11NO3 |
Molecular Weight (g/mol) | 193.20 |
MDL Number | MFCD00044399 |
InChI Key | YKAKNMHEIJUKEX-UHFFFAOYSA-N |
Synonym | 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid |
PubChem CID | 99223 |
ChEBI | CHEBI:68500 |
SMILES | CC1=CC=CC(=C1)C(=O)NCC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15490288
|
Thermo Scientific Alfa Aesar
H53458.03 |
1 g |
532.80 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400298
|
Thermo Scientific Alfa Aesar
H53458.06 |
5 g |
1836.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15410298
|
Thermo Scientific Alfa Aesar
H53458.14 |
25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27115-49-7 | |
193.20 | |
YKAKNMHEIJUKEX-UHFFFAOYSA-N | |
99223 | |
CC1=CC=CC(=C1)C(=O)NCC(O)=O |
C10H11NO3 | |
MFCD00044399 | |
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid | |
CHEBI:68500 |
Specifications
27115-49-7 | |
C10H11NO3 | |
3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid | |
CC1=CC=CC(=C1)C(=O)NCC(O)=O | |
193.20 | |
CHEBI:68500 | |
97% |
138°C to 140°C | |
MFCD00044399 | |
YKAKNMHEIJUKEX-UHFFFAOYSA-N | |
2-[(3-methylphenyl)formamido]acetic acid | |
99223 | |
193.2 | |
3-Methylhippuric acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only