missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhippuric acid, 97%, Thermo Scientific Chemicals

Product Code. 15400298
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27115-49-7
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00044399
InChI Key YKAKNMHEIJUKEX-UHFFFAOYSA-N
Synonym 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
PubChem CID 99223
ChEBI CHEBI:68500
SMILES CC1=CC=CC(=C1)C(=O)NCC(O)=O

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
IUPAC Name 2-[(3-methylbenzoyl)amino]acetic acid
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 3-Methylhippuric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'