missing translation for 'onlineSavingsMsg'
Learn More
3-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11414514

3-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11414514
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11414514

Brand: Thermo Scientific Alfa Aesar A13140.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-27-9
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042286
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchlorideShow MoreShow Less
PubChem CID 2734258
IUPAC Name 3-fluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 7°C
Density 1.463
Boiling Point 231°C to 232°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 5 g
UN Number UN3265
Beilstein 2091631
Sensitivity Moisture sensitive
Formula Weight 194.61
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzenesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ1452A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13140.06,A13140.03
Lot NumberF01P38 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13140.06,A13140.03
Lot Number10236141 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13140.06,A13140.03
Lot Number10109228 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13140.06,A13140.03
Lot NumberB15M32 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13140.06,A13140.03
compliance-icons
Product Identifier
  • 3-Fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : 3-Fluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.