Learn More
3-Fluorobenzenesulfonyl chloride, 97% Product Code.: 11404514
3-Fluorobenzenesulfonyl chloride, 97%
Product Code. 11404514
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11404514
Brand: Thermo Scientific Alfa Aesar A13140.03
Request Bulk or Custom Format
CAS: 701-27-9 | C6H4ClFO2S | 194.60 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 701-27-9
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042286
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
PubChem CID 2734258
IUPAC Name 3-fluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC=C1)S(Cl)(=O)=O
Specifications
Melting Point 7°C
Density 1.463
Boiling Point 231°C to 232°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
UN Number UN3265
Beilstein 2091631
Sensitivity Moisture sensitive
Formula Weight 194.61
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzenesulfonyl chloride
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberJ1452A Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13140.06,A13140.03
Lot NumberF01P38 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13140.06,A13140.03
Lot Number10236141 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13140.06,A13140.03
Lot Number10109228 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13140.06,A13140.03
Lot NumberH1320A Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA13140.06,A13140.03
Product Title
3-Fluorobenzenesulfonyl chloride, 97% >