Learn More
3-Chloro-4-methylphenyl isocyanate, 98%, Thermo Scientific Chemicals
309.00 NOK - 2580.00 NOK
Chemical Identifiers
CAS | 28479-22-3 |
---|---|
Molecular Formula | C8H6ClNO |
Molecular Weight (g/mol) | 167.59 |
MDL Number | MFCD00013858 |
InChI Key | UKTKKMZDESVUEE-UHFFFAOYSA-N |
Synonym | 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison |
PubChem CID | 62832 |
IUPAC Name | 2-chloro-4-isocyanato-1-methylbenzene |
SMILES | CC1=CC=C(C=C1Cl)N=C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338595
|
Thermo Scientific Alfa Aesar
L12064.03 |
1 g |
309.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348595
|
Thermo Scientific Alfa Aesar
L12064.06 |
5 g |
679.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358595
|
Thermo Scientific Alfa Aesar
L12064.14 |
25 g |
2580.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28479-22-3 | |
167.59 | |
UKTKKMZDESVUEE-UHFFFAOYSA-N | |
62832 | |
CC1=CC=C(C=C1Cl)N=C=O |
C8H6ClNO | |
MFCD00013858 | |
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison | |
2-chloro-4-isocyanato-1-methylbenzene |
Specifications
20°C to 24°C | |
1.224 | |
1.556 | |
MFCD00013858 | |
2690771 | |
UKTKKMZDESVUEE-UHFFFAOYSA-N | |
2-chloro-4-isocyanato-1-methylbenzene | |
62832 | |
98% |
28479-22-3 | |
225°C | |
C8H6ClNO | |
UN2236 | |
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison | |
CC1=CC=C(C=C1Cl)N=C=O | |
167.59 | |
167.6 | |
3-Chloro-4-methylphenyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H314-H318
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P284-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H330-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-CHLORO-4-METHYLPHENYL ISOCYANATE, LIQUID
missing translation for 'einecsNumber' : 249-050-7
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- 3-Chloro-4-methylphenyl isocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 3-Chloro-4-methylphenyl isocyanate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.