missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenyl isocyanate, 98%

Product Code. 11348595
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 28479-22-3 | C8H6ClNO | 167.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28479-22-3
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00013858
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
PubChem CID 62832
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Melting Point 20°C to 24°C
Density 1.224
Boiling Point 225°C
Quantity 5 g
Refractive Index 1.556
UN Number UN2236
Beilstein 2690771
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-4-methylphenyl isocyanate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.