Learn More
2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals
264.75 NOK - 1075.00 NOK
Chemical Identifiers
CAS | 5653-40-7 |
---|---|
Molecular Formula | C9H11NO4 |
Molecular Weight (g/mol) | 197.19 |
MDL Number | MFCD00011671 |
InChI Key | HJVAVGOPTDJYOJ-UHFFFAOYSA-N |
Synonym | 4,5-dimethoxyanthranilic acid, 6-aminoveratric acid, benzoic acid, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy benzoic acid, 2-amino-4,5-dimethoxy-benzoic acid, 6-aminoveratricacid, zlchem 238, pubchem3110, acmc-1b03r, ksc269i1n |
PubChem CID | 79736 |
IUPAC Name | 2-amino-4,5-dimethoxybenzoic acid |
SMILES | COC1=CC(N)=C(C=C1OC)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322416
|
Thermo Scientific Alfa Aesar
L06205.06 |
5 g | N/A | ||||||
11332416
|
Thermo Scientific Alfa Aesar
L06205.14 |
25 g |
1075.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in Methanol and Water
Notes
Incompatible with bases and oxidizing agents. Store at -20°C.
Chemical Identifiers
5653-40-7 | |
197.19 | |
HJVAVGOPTDJYOJ-UHFFFAOYSA-N | |
79736 | |
COC1=CC(N)=C(C=C1OC)C(O)=O |
C9H11NO4 | |
MFCD00011671 | |
4,5-dimethoxyanthranilic acid, 6-aminoveratric acid, benzoic acid, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy benzoic acid, 2-amino-4,5-dimethoxy-benzoic acid, 6-aminoveratricacid, zlchem 238, pubchem3110, acmc-1b03r, ksc269i1n | |
2-amino-4,5-dimethoxybenzoic acid |
Specifications
5653-40-7 | |
C9H11NO4 | |
782814 | |
Soluble in Methanol and Water | |
COC1=CC(N)=C(C=C1OC)C(O)=O | |
197.19 | |
197.19 | |
2-Amino-4,5-dimethoxybenzoic acid |
∽155°C to 158°C (decomposition) | |
MFCD00011671 | |
4,5-dimethoxyanthranilic acid, 6-aminoveratric acid, benzoic acid, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy benzoic acid, 2-amino-4,5-dimethoxy-benzoic acid, 6-aminoveratricacid, zlchem 238, pubchem3110, acmc-1b03r, ksc269i1n | |
HJVAVGOPTDJYOJ-UHFFFAOYSA-N | |
2-amino-4,5-dimethoxybenzoic acid | |
79736 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 227-095-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures