missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11322416
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11322416

Brand: Thermo Scientific Alfa Aesar L06205.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4,5-dimethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Methanol and Water

Notes
Incompatible with bases and oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5653-40-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00011671
InChI Key HJVAVGOPTDJYOJ-UHFFFAOYSA-N
Synonym 4,5-dimethoxyanthranilic acid, 6-aminoveratric acid, benzoic acid, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy benzoic acid, 2-amino-4,5-dimethoxy-benzoic acid, 6-aminoveratricacid, zlchem 238, pubchem3110, acmc-1b03r, ksc269i1n
PubChem CID 79736
IUPAC Name 2-amino-4,5-dimethoxybenzoic acid
SMILES COC1=CC(N)=C(C=C1OC)C(O)=O

Specifications

Melting Point ∽155°C to 158°C (decomposition)
Quantity 5 g
Beilstein 782814
Solubility Information Soluble in Methanol and Water
Formula Weight 197.19
Percent Purity 98%
Chemical Name or Material 2-Amino-4,5-dimethoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.