missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 18mm PP Screw Cap, Black, Center hole, 18-400 thread
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 18 08 0516
Description
- 18mm PP Screw cap
- Black
- Closed top

Specifications
Black | |
Polypropylene | |
Center Hole Screw Cap |
18 mm | |
100 | |
ND18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ 18mm PP Screw Cap, Black, Center hole, 18-400 thread > centre hole,black
Spot an opportunity for improvement?Share a Content Correction