Learn More
Bochem™ 18/10 Stainless Steel Rods
Brand:  Bochem™ 5182
 View more versions of this product
Product Code. 10204561
  • 344.00 NOK / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Specifications
Stainless Steel (18/10)
Rod
Stand bases
750 mm
For Stand Bases
1 Ea.
Product Title
Bochem™ 18/10 Stainless Steel Rods > Diameter: 13mm; Length: 750mm; For Use With Stand Bases