Learn More
ONO-7300243, MedChemExpress
MedChemExpress ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.
Brand: Medchem Express HY-100882
Description
ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.Specifications
ONO-7300243 | |
White | |
C28H31NO5 | |
DMSO : 100 mg/mL (216.66 mM; Need ultrasonic) | |
461.55 | |
98.14% | |
Solid |
638132-34-0 | |
Research | |
50 mg | |
O=C(O)CC1=CC=C(CN(C(C2=CC(OC)=C(C)C(OC)=C2)=O)CCCC3=CC=CC=C3)C=C1 | |
461.55 | |
Research | |
COVID-19-immunoregulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only