missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Double Spatula, Flexible

Use Bochem™ 18/10 Stainless Steel Double Spatula, Flexible for your laboratory needs..

Brand:  Bochem™ 3624

 View more versions of this product

Product Code. 10346241

  • 71.90 NOK / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 18/10 Stainless Steel
  • Flexible
Specifications

Specifications

1 Ea.
Straight
50 mm
Stainless Steel (18/10)
250 x 10 mm
250 mm
Autoclavable
Double Ended Spatula
10 mm
4mm
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers