Learn More
1-Naphthyl PP1 hydrochloride, MedChemExpress
MedChemExpress 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
Brand:  Medchem Express HY-13941B
Encompass_Preferred
 View more versions of this product
Product Code. 16482067
  • 6700.78 NOK / 50mg
Description
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
Specifications
Specifications
1-Naphthyl PP1 hydrochloride
956025-47-1
50 mg
C19H20ClN5
1-NA-PP 1 hydrochloride
CC(N1N=C(C2=C3C=CC=CC3=CC=C2)C4=C(N)N=CN=C41)(C)C.[H]Cl
353.85
Research
Solid
White
Research
Cancer-Kinase/protease
DMSO : ≥ 73 mg/mL (206.30 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
353.85
99.94%
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
1-Naphthyl PP1 hydrochloride, MedChemExpress