Learn More
1-Naphthyl PP1 hydrochloride, MedChemExpress
MedChemExpress 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.

1311.71 NOK - 10778.14 NOK
Specifications
Chemical Name or Material | 1-Naphthyl PP1 hydrochloride |
---|---|
Physical Form | Solid |
CAS | 956025-47-1 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16495918
![]() |
Medchem Express
HY-13941B |
10 mg |
2086.09 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16482067
![]() |
Medchem Express
HY-13941B |
50 mg |
6700.78 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468557
![]() |
Medchem Express
HY-13941B |
5 mg |
1311.71 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436366
![]() |
Medchem Express
HY-13941B |
100 mg |
10778.14 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.Specifications
1-Naphthyl PP1 hydrochloride | |
956025-47-1 | |
Research | |
Cancer-Kinase/protease | |
DMSO : ≥ 73 mg/mL (206.30 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) | |
353.85 | |
99.94% |
Solid | |
White | |
C19H20ClN5 | |
1-NA-PP 1 hydrochloride | |
CC(N1N=C(C2=C3C=CC=CC3=CC=C2)C4=C(N)N=CN=C41)(C)C.[H]Cl | |
353.85 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.