Learn More
1-Naphthyl PP1 hydrochloride, MedChemExpress
MedChemExpress 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
Brand: Medchem Express HY-13941B

Description
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.Specifications
1-Naphthyl PP1 hydrochloride | |
956025-47-1 | |
100 mg | |
C19H20ClN5 | |
1-NA-PP 1 hydrochloride | |
CC(N1N=C(C2=C3C=CC=CC3=CC=C2)C4=C(N)N=CN=C41)(C)C.[H]Cl | |
353.85 | |
Research |
Solid | |
White | |
Research | |
Cancer-Kinase/protease | |
DMSO : ≥ 73 mg/mL (206.30 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) | |
353.85 | |
99.94% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only