missing translation for 'onlineSavingsMsg'
Learn More
1,7-Octadiene, 98.5%, Thermo Scientific Chemicals Product Code.: 10075683

1,7-Octadiene, 98.5%, Thermo Scientific Chemicals

Product Code. 10075683
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 10075683

Brand: Thermo Scientific Acros 129355000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 114.0°C to 121.0°C
Flash Point 9°C
Infrared Spectrum Authentic
Assay Percent Range 98.50%
Packaging Glass bottle
Linear Formula H2C=CH(CH2)4CH=CH2
Refractive Index 1.4210 to 1.4230
Quantity 500 mL
UN Number 2309
Specific Gravity 0.74
Solubility Information Solubility in water: 0.08g/L (20°C). Other solubilities: miscible with methanol, cyclohexane, benzene,, isopropanol and petroleumether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 110.2
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1, 7-Octadiene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462199 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number129355000,129351000
Lot NumberA0454803 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number129355000,129351000
Lot NumberA0452507 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number129355000,129351000
Lot NumberA0402994 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number129355000,129351000
Lot NumberA0402354 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number129355000,129351000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,7-Octadiene, 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.