missing translation for 'onlineSavingsMsg'
Learn More
1,7-Octadiene, 97%, Thermo Scientific Chemicals Product Code.: 11362007

1,7-Octadiene, 97%, Thermo Scientific Chemicals

Product Code. 11362007
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11362007

Brand: Thermo Scientific Alfa Aesar L07659.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3710-30-3 | C8H14 | 110.20 g/mol

1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

Solubility
Miscible with methanol and cyclohexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1Show MoreShow Less
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

Melting Point <-70°C
Density 0.735
Boiling Point 118°C to 120°C
Flash Point 5°C (41°F)
Odor Strong
Linear Formula CH2=CH(CH2)4CH=CH2
Refractive Index 1.422
Quantity 500 mL
UN Number UN2309
Beilstein 605288
Solubility Information Miscible with methanol and cyclohexane. Slightly miscible with water.
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material 1,7-Octadiene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462199 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberL07659.AE,L07659.AP
Lot NumberA0454803 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog NumberL07659.AE,L07659.AP
Lot NumberA0452507 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberL07659.AE,L07659.AP
Lot Number10207158 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07659.AE,L07659.AP
Lot Number10177609 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL07659.AE,L07659.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,7-Octadiene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.